Skip to Header
Skip to Main Content
Skip to Footer
Avansert
Avansert
SDS
Sertifikat
Produsentdata
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Chloro-4-methoxy-5-nitropyridine
2-Chloro-4-methoxy-5-nitropyridine
2-Chloro-4-methoxy-5-nitropyridine
Irritant
2-Chloro-4-methoxy-5-nitropyridine
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2-Chloro-4-methoxy-5-nitropyridine
Supplier:
Apollo Scientific
About this item
CAS Number:
607373-83-1
:
Formula:
C₆H₅ClN₂O₃
MDL Number:
MFCD09907961
:
188.569 g/mol
:
GHS07
Storage Temperature:
Romtemperatur