Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Logg inn
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Amidinobenzoic acid hydrochloride
4-Amidinobenzoic acid hydrochloride 97%
4-Amidinobenzoic acid hydrochloride 97%
Irritant
4-Amidinobenzoic acid hydrochloride 97%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
4-Amidinobenzoic acid hydrochloride 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
42823-72-3
:
Formula:
C₈H₈N₂O₂·ClH
MDL Number:
MFCD00269643
:
200.622 g/mol
:
GHS07
Storage Temperature:
Ambient