7-Chloro-1,8-naphthyridin-2-amine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:15944-33-9
:
Formula:C₈H₆ClN₃
MDL Number:MFCD11878574
:179.606 g/mol
Melting Pt:186…188 °C
EINECS:240-078-5
:GHS07
Storage Temperature:Romtemperatur