Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
7-Bromo-5H-pyrido[4,3-b]indole
7-Bromo-5H-pyrido[4,3-b]indole 95+%
7-Bromo-5H-pyrido[4,3-b]indole 95+%
Irritant
7-Bromo-5H-pyrido[4,3-b]indole 95+%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
7-Bromo-5H-pyrido[4,3-b]indole 95+%
Supplier:
Apollo Scientific
About this item
CAS Number:
1015460-59-9
:
Formula:
C₁₁H₇BrN₂
MDL Number:
MFCD18803689
:
247.091 g/mol
:
GHS07
Storage Temperature:
Refrigerator