Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,4-Dihydroxy-6-methylbenzaldehyde
2,4-Dihydroxy-6-methylbenzaldehyde
2,4-Dihydroxy-6-methylbenzaldehyde
Irritant
2,4-Dihydroxy-6-methylbenzaldehyde
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2,4-Dihydroxy-6-methylbenzaldehyde
Supplier:
Apollo Scientific
About this item
CAS Number:
487-69-4
:
Formula:
C₈H₈O₃
MDL Number:
MFCD00033852
:
152.147 g/mol
Melting Pt:
180…184 °C
:
GHS07
Storage Temperature:
Ambient