Order Entry
Norway
ContactUsLinkComponent
(S)-BI-DIME ≥98%
(S)-BI-DIME ≥98%
Catalog # APOSOR510254-100MG
Supplier:  Apollo Scientific
CAS Number:  
(S)-BI-DIME ≥98%
Catalog # APOSOR510254-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :330.358 g/mol
Storage Temperature:Ambient
CAS Number:1373432-09-7
 :MFCD31630893
Formula:C₁₉H₂₃O₃P
MDL Number:MFCD31630704