Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Iodo-2-methylbenzaldehyde
3-Iodo-2-methylbenzaldehyde 96%
3-Iodo-2-methylbenzaldehyde 96%
3-Iodo-2-methylbenzaldehyde 96%
Supplier:
Apollo Scientific
Certificates
Expand image
3-Iodo-2-methylbenzaldehyde 96%
Supplier:
Apollo Scientific
About this item
CAS Number:
848444-83-7
:
Formula:
C₈H₇IO
MDL Number:
MFCD18397774
:
246.045 g/mol
:
GHS07
Storage Temperature:
Ambient