N-(2-Chlorophenyl)cinnamamide
Catalog # APOSOR470382-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:257.715 g/mol
Storage Temperature:Ambient
CAS Number:73108-79-9
Formula:C₁₅H₁₂ClNO
MDL Number:MFCD00157758