Order Entry
Norway
ContactUsLinkComponent
 
(S)-N-Cbz-2-amino-1-phenylethylamine 97%
Catalog # APOSOR452159-1G
Supplier:  Apollo Scientific
CAS Number:  
(S)-N-Cbz-2-amino-1-phenylethylamine 97%
Catalog # APOSOR452159-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :270.326 g/mol
Storage Temperature:Ambient
CAS Number:130406-36-9
Formula:C₁₆H₁₈N₂O₂
MDL Number:MFCD09260584