N-(3-Azetidinyl)piperidine dihydrochloride
Catalog # APOSOR451002-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:213.148 g/mol
Storage Temperature:Ambient
CAS Number:178312-57-7
Formula:C₈H₁₆N₂·2ClH
MDL Number:MFCD11617822