Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 3-bromo-5-formylbenzoate
Methyl 3-bromo-5-formylbenzoate
Methyl 3-bromo-5-formylbenzoate
Irritant
Methyl 3-bromo-5-formylbenzoate
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Methyl 3-bromo-5-formylbenzoate
Supplier:
Apollo Scientific
About this item
CAS Number:
377734-27-5
:
Formula:
C₉H₇BrO₃
MDL Number:
MFCD22381229
:
243.054 g/mol
:
GHS07
Storage Temperature:
Ambient