kr 0,00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Norway
ContactUsLinkComponent
Methyl 5-bromo-2-formylbenzoate
Methyl 5-bromo-2-formylbenzoate
Methyl 5-bromo-2-formylbenzoate
:
Apollo Scientific
Methyl 5-bromo-2-formylbenzoate
:
Apollo Scientific
:
1016163-89-5
:
:
C₉H₇BrO₃
:
MFCD16036575
:
243.054 g/mol
:
GHS07
:
Ambient