Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Bromo-3-formylbenzoic acid
2-Bromo-3-formylbenzoic acid
2-Bromo-3-formylbenzoic acid
:
Apollo Scientific
2-Bromo-3-formylbenzoic acid
:
Apollo Scientific
:
1289208-00-9
:
:
C₈H₅BrO₃
:
MFCD28357159
:
229.027 g/mol
:
GHS07
:
Ambient