Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Bromo-4-nitrobenzenesulphonyl chloride
2-Bromo-4-nitrobenzenesulphonyl chloride
2-Bromo-4-nitrobenzenesulphonyl chloride
Corrosive
2-Bromo-4-nitrobenzenesulphonyl chloride
Supplier:
Apollo Scientific
Certificates
Expand image
Corrosive
2-Bromo-4-nitrobenzenesulphonyl chloride
Supplier:
Apollo Scientific
About this item
CAS Number:
1099660-60-2
:
Formula:
C₆H₃BrClNO₄S
MDL Number:
MFCD11650638
:
300.514 g/mol
Storage Temperature:
Ambient
UN:
3261
ADR:
8,III
:
GHS05