Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-(2,5-Dichlorophenyl)-1H-pyrazole
3-(2,5-Dichlorophenyl)-1H-pyrazole
3-(2,5-Dichlorophenyl)-1H-pyrazole
Corrosive
Irritant
3-(2,5-Dichlorophenyl)-1H-pyrazole
Supplier:
Apollo Scientific
Certificates
Expand image
Corrosive
Irritant
3-(2,5-Dichlorophenyl)-1H-pyrazole
Supplier:
Apollo Scientific
About this item
CAS Number:
59843-71-9
:
Formula:
C₉H₆Cl₂N₂
MDL Number:
MFCD02091525
:
213.063 g/mol
:
GHS05
:
GHS07
Storage Temperature:
Ambient