Order Entry
Norway
ContactUsLinkComponent
Methyl 5-chloro-1-methyl-3-phenyl-1H-pyrazole-4-carboxylate, tech
Methyl 5-chloro-1-methyl-3-phenyl-1H-pyrazole-4-carboxylate, tech
Catalog # APOSOR32862-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl 5-chloro-1-methyl-3-phenyl-1H-pyrazole-4-carboxylate, tech
Catalog # APOSOR32862-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :250.681 g/mol
Storage Temperature:Ambient
CAS Number:956764-06-0
Formula:C₁₂H₁₁ClN₂O₂
MDL Number:MFCD06496183