4-Bromo-3-[(ethylamino)methyl]phenol, tech
Catalog # APOSOR32804-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:230.102 g/mol
Storage Temperature:Ambient
CAS Number:1281872-64-7
Formula:C₉H₁₂BrNO
MDL Number:MFCD18904323