Order Entry
Norway
ContactUsLinkComponent
Methyl 1-oxo-2-[(pyridin-2-yl)methyl]-1,2-dihydroisoquinoline-4-carboxylate, tech
Methyl 1-oxo-2-[(pyridin-2-yl)methyl]-1,2-dihydroisoquinoline-4-carboxylate, tech
Catalog # APOSOR32286-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl 1-oxo-2-[(pyridin-2-yl)methyl]-1,2-dihydroisoquinoline-4-carboxylate, tech
Catalog # APOSOR32286-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :294.305 g/mol
Storage Temperature:Ambient
CAS Number:303995-32-6
Formula:C₁₇H₁₄N₂O₃
MDL Number:MFCD00140339