Order Entry
Norway
ContactUsLinkComponent
Ethyl 5-(4-chlorophenyl)-1-methyl-1H-pyrazole-3-carboxylate
Ethyl 5-(4-chlorophenyl)-1-methyl-1H-pyrazole-3-carboxylate
Catalog # APOSOR310530-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 5-(4-chlorophenyl)-1-methyl-1H-pyrazole-3-carboxylate
Catalog # APOSOR310530-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :264.708 g/mol
Storage Temperature:Ambient
CAS Number:864426-88-0
Formula:C₁₃H₁₃ClN₂O₂
MDL Number:MFCD08446156