kr 0,00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Norway
ContactUsLinkComponent
2-Bromo-5-(3-nitrophenoxy)-1,3,4-thiadiazole
2-Bromo-5-(3-nitrophenoxy)-1,3,4-thiadiazole
2-Bromo-5-(3-nitrophenoxy)-1,3,4-thiadiazole
2-Bromo-5-(3-nitrophenoxy)-1,3,4-thiadiazole
Supplier:
Apollo Scientific
Certificates
Expand image
2-Bromo-5-(3-nitrophenoxy)-1,3,4-thiadiazole
Supplier:
Apollo Scientific
About this item
CAS Number:
1339381-69-9
:
Formula:
C₈H₄BrN₃O₃S
MDL Number:
MFCD20368359
:
302.105 g/mol
Melting Pt:
111…113 °C
Storage Temperature:
Ambient