5-Oxaspiro[3.5]nonan-8-ol
Catalog # APOSOR304415-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:142.196 g/mol
Storage Temperature:Ambient
CAS Number:1368181-23-0
Formula:C₈H₁₄O₂
MDL Number:MFCD22070788