Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-tert-Butyl-1H-imidazole-4-carbaldehyde
5-tert-Butyl-1H-imidazole-4-carbaldehyde 95%
5-tert-Butyl-1H-imidazole-4-carbaldehyde 95%
Irritant
5-tert-Butyl-1H-imidazole-4-carbaldehyde 95%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
5-tert-Butyl-1H-imidazole-4-carbaldehyde 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
714273-83-3
:
Formula:
C₈H₁₂N₂O
MDL Number:
MFCD14583325
:
152.194 g/mol
Melting Pt:
122…124 °C
:
GHS07
Storage Temperature:
Ambient