Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Bromo-5-methylpicolinaldehyde
4-Bromo-5-methylpicolinaldehyde 97%
4-Bromo-5-methylpicolinaldehyde 97%
APOSOR302618-250MG
:
Apollo Scientific
:
4-Bromo-5-methylpicolinaldehyde 97%
APOSOR302618-250MG
:
Apollo Scientific
:
Pakketype:
Bottle
Fp:
250 mg
Seq:
2590
:
:
200.033 g/mol
:
Romtemperatur
:
1196157-14-8
:
C₇H₆BrNO
:
MFCD13190304