
Ethyl 6-cyanonicotinate 97+%
Catalog # APOSOR301302-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:176.172 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:76196-79-7
Formula:C₉H₈N₂O₂
MDL Number:MFCD06637534