Order Entry
Norway
ContactUsLinkComponent
1-(4,5-Dihydro-1H-imidazol-2-yl)-3,5-dimethyl-1H-pyrazole hydrobromide
Irritant
1-(4,5-Dihydro-1H-imidazol-2-yl)-3,5-dimethyl-1H-pyrazole hydrobromide
Catalog # APOSOR23795-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1-(4,5-Dihydro-1H-imidazol-2-yl)-3,5-dimethyl-1H-pyrazole hydrobromide
Catalog # APOSOR23795-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :245.12 g/mol
Melting Pt:270…275 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:132369-02-9
Formula:C₈H₁₂N₄·BrH
MDL Number:MFCD00053026