
1-[4-(Bromomethyl)phenyl]-1H-1,2,4-triazole hydrobromide hydrate
Catalog # APOSOR23306-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:337.011 g/mol
Melting Pt:280 °C
Storage Temperature:Ambient
UN:1759
ADR:8,III
:GHS05
CAS Number:1138011-23-0
Formula:C₉H₈BrN₃·BrH·H₂O
MDL Number:MFCD17015228