Ethyl 1-(4-chlorophenyl)-5-phenyl-4,5-dihydro-1H-pyrazole-3-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
:
Formula:C₁₈H₁₇ClN₂O₂
MDL Number:MFCD29918605
:328.793 g/mol
Storage Temperature:Ambient
Frequently Bought Together