L-Thyroxine-[L-Tyr-ring-13C6] hydrochloride
Catalog # APOSBISC1052-1MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 mg
- Seq:2070
Specifications
About this item
:
:819.287 g/mol
Storage Temperature:Ambient
Formula:[₁₃C]₆C₉H₁₁I₄NO₄·ClH
MDL Number:MFCD11656670