Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Hydroxy-PEG1-t-butyl ester
Hydroxy-PEG1-t-butyl ester
Hydroxy-PEG1-t-butyl ester
Irritant
Hydroxy-PEG1-t-butyl ester
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Hydroxy-PEG1-t-butyl ester
Supplier:
Apollo Scientific
About this item
CAS Number:
671802-00-9
:
Formula:
C₉H₁₈O₄
MDL Number:
MFCD22056310
:
190.237 g/mol
:
GHS07
Storage Temperature:
Ambient