Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Azido-PEG12-acid
Azido-PEG12-acid
Azido-PEG12-acid
Irritant
Azido-PEG12-acid
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Azido-PEG12-acid
Supplier:
Apollo Scientific
About this item
CAS Number:
2152679-71-3
:
1167575-20-3
Formula:
C₂₇H₅₃N₃O₁₄
MDL Number:
MFCD21363301
:
643.721 g/mol
:
GHS07
Storage Temperature:
Refrigerator