kr 0,00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Norway
ContactUsLinkComponent
Amino-PEG5-t-butyl ester
Amino-PEG5-t-butyl ester
Amino-PEG5-t-butyl ester
:
Apollo Scientific
Amino-PEG5-t-butyl ester
:
Apollo Scientific
:
1446282-18-3
:
:
C₁₇H₃₅NO₇
:
MFCD25424125
:
365.462 g/mol
:
GHS07
:
Refrigerator