Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Amino-PEG12-acid
Amino-PEG12-Acid
Amino-PEG12-Acid
Irritant
Amino-PEG12-Acid
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Amino-PEG12-Acid
Supplier:
Apollo Scientific
About this item
CAS Number:
196936-04-6
:
663921-15-1
Formula:
C₂₇H₅₅NO₁₄
MDL Number:
MFCD11041150
:
617.724 g/mol
:
GHS07
Storage Temperature:
Ambient