Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Amino-PEG5-acid
Amino-PEG5-acid
Amino-PEG5-acid
Amino-PEG5-acid
Supplier:
Apollo Scientific
Certificates
Expand image
Amino-PEG5-acid
Supplier:
Apollo Scientific
About this item
CAS Number:
1191078-74-6
:
Formula:
C₁₃H₂₇NO₇
MDL Number:
MFCD25424126
:
309.356 g/mol
Storage Temperature:
Ambient