Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Chloro-8-hydroxyquinoline sulphate
5-Chloro-8-hydroxyquinoline sulphate ≥95%
5-Chloro-8-hydroxyquinoline sulphate ≥95%
Irritant
5-Chloro-8-hydroxyquinoline sulphate ≥95%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
5-Chloro-8-hydroxyquinoline sulphate ≥95%
Supplier:
Apollo Scientific
About this item
CAS Number:
15164-40-6
:
MDL Number:
MFCD28122834
Melting Pt:
186…188 °C
:
GHS07
Storage Temperature:
Freezer