Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Carbenicillin disodium salt
Carbenicillin disodium salt
Carbenicillin disodium salt
Carbenicillin disodium salt
Supplier:
Apollo Scientific
Certificates
Utvid bildet
Carbenicillin disodium salt
Supplier:
Apollo Scientific
About this item
CAS Number:
4800-94-6
:
Formula:
C₁₇H₁₆N₂O₆S·2Na
MDL Number:
MFCD00077683
:
422.363 g/mol
EINECS:
225-360-8
:
GHS07
:
GHS08
Storage Temperature:
Refrigerator