Order Entry
Norway
ContactUsLinkComponent
 
Polyvinyl alcohol, GPR RECTAPUR®
Catalog # 87101.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
Polyvinyl alcohol, GPR RECTAPUR®
Catalog # 87101.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    HDPE bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Appearance (colorless translucent granules):
    Passes test
  • Degree of hydrolysis:
    98 - 99%
  • Degree of polymerization:
    1700 - 1800
  • pH (0,2% in water):
    5,0 - 7,0
  • Viscosity (25°C):
    25 - 32 mPas
  • Ash content:
    max. 0,7%
  • Volatile matter:
    max. 0,5%
  • Cl (Chloride):
    max. 10 ppm

Specification Test Results

About this item

 :
CAS Number:9002-89-5
 :25213-24-5
Formula:[-CH₂CH(OH)-]n
MDL Number:MFCD00081922