Order Entry
Norway
ContactUsLinkComponent
Collodion 2% in amyl acetate for microscopy, Sigma-Aldrich®
Flammable
Collodion 2% in amyl acetate for microscopy, Sigma-Aldrich®
Catalog # 09817-100ML
CAS Number:  
Flammable
Collodion 2% in amyl acetate for microscopy, Sigma-Aldrich®
Catalog # 09817-100ML
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
ADR:3,I
 :GHS02
CAS Number:9004-70-0
MDL Number:MFCD00081525