Order Entry
Norway
ContactUsLinkComponent
Lucifer Yellow VS dilithium salt ~85%, Sigma-Aldrich®
Lucifer Yellow VS dilithium salt ~85%, Sigma-Aldrich®
Certificates
undefined
Lucifer Yellow VS dilithium salt ~85%, Sigma-Aldrich®

Specification Test Results

  • Appearance (Colour):
    Yellow to Brown
  • Appearance (Form):
    Powder
  • Infrared spectrum:
    Conforms to Structure
  • Solubility (Colour):
    Light Yellow to Brown
  • Solubility (Turbidity):
    Clear to Slightly Hazy
  • Purity (HPLC):
    ≥80.00 %

Specification Test Results