SynaptoGreen™ C4 ≥95% (by HPLC), Sigma-Aldrich®
Catalog # S6814-5MG
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:5 mg
- Seq:2210
Specifications
About this item
:
:611.54 g/mol
Storage Temperature:Freezer
CAS Number:149838-22-2
Formula:C₃₀H₄₉Br₂N₃
MDL Number:MFCD00467815