Order Entry
Norway
ContactUsLinkComponent
Coproporphyrin I tetramethyl ester ≥90% (by HPLC), Sigma-Aldrich®
Coproporphyrin I tetramethyl ester ≥90% (by HPLC), Sigma-Aldrich®
  C4529-1MG
 :  
undefined
Coproporphyrin I tetramethyl ester ≥90% (by HPLC), Sigma-Aldrich®
  C4529-1MG
 :  

 

  • Pk:
    1 mg
  • Seq:
    2070

 

 

 :
 :252…254 °C (lit.)
 :Ambient
 :25767-20-8
 :MFCD00079038