Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Chlorazol black E
Chlorazol black E, Sigma-Aldrich®
Chlorazol black E, Sigma-Aldrich®
Health
Irritant
Chlorazol black E, Sigma-Aldrich®
Supplier:
SIGMA ALDRICH MICROSCOPY
Certificates
Expand image
Health
Irritant
Chlorazol black E, Sigma-Aldrich®
Supplier:
SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:
1937-37-7
:
Formula:
C₃₄H₂₇N₉O₇S₂
MDL Number:
MFCD00066363
:
781.73 g/mol
:
GHS08
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
Documentation