Bacteriochlorophyll from Rhodopseudomonas sphaeroides, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:17499-98-8
:
Formula:C₅₅H₇₄MgN₄O₆
MDL Number:MFCD00078976
:911.5 g/mol
:M-Clarity™ quality level = MQ200
Storage Temperature:Freezer
Specification Test Results
- Solubility (Form)
10 mg/mL, EtOH:Solution - Magnesium (Mg):1.5 - 3.0 %
- E1%
in Ethyl Ether:150 - 250 - Lambda Max:570 - 578 nm
- Note:Absorbance maxima are consistent with Bacteriochlorophyll.
Specification Test Results
Frequently Bought Together