Order Entry
Norway
Orders LinkContactUsLinkComponent
Hydrogen peroxide 12% (w/v)

Hydrogen peroxide 12% (w/v)

Oxidizing
Corrosive
Hydrogen peroxide 12% (w/v)
Certificates
Hydrogen peroxide 12% (w/v)

2 Options Available Below

Product Details & Documents
  • Reagent for chemical synthesis
  • Oxidising reagent
  • CAS Number:7722-84-1
  • Formula:H₂O₂
  • MDL Number:MFCD00011333
  • Molecular Weight:34.01 g/mol
  • Boiling Pt:100 °C (760 mmHg)
  • Density:1.0500 g/ml
  • Storage Temperature:Ambient
  • UN:2984
  • ADR:5.1,III
  •  :Density: 1.11 g/cm³ (20 °C)

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear colorless Solution
  • Titration with KMnO4:
    11.0 to 13.0 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Plastic bottle
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - kr 325,00
    kr 325,00
    1 * 100 mL
       kr 325,00

  • Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - kr 625,00
    kr 625,00
    1 * 1 L
       kr 625,00

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.