Bromocresol purple, AnalaR NORMAPUR® ACS
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:115-40-2
:
Formula:C₂₁H₁₆Br₂O₅S
MDL Number:MFCD00011681
:540.23 g/mol
Specification Test Results
- Clarity of solution:Passes test ACS
- Identification (UV/VIS):Passes test
- Sensitivity:Passes test
- Visual transition interval:pH 5.2 - pH 6.8
- Loss on drying (110°C):Max. 1 %
- Absorption max. λ 1 max (buffer pH 5.2):429 - 433 nm
- Absorption max. λ 2 max (buffer pH 6.8):587 - 590 nm
- Sp.absorptivity (λ1max; 0.005g/l; pH5.2; dry):400 - 470
- Sp.absorptivity (λ2max; 0.005g/l; pH6.8; dry):1000 - 1160
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_VWRC85723.050 spec_VWRC85723.050 spec_VWRC85723.130 spec_VWRC85723.130