Order Entry
Norway
ContactUsLinkComponent
Water purification systems, Milli-Q® IQ 7003/05/10/15
Water purification systems, Milli-Q® IQ 7003/05/10/15
Leverandør:  Merck
Sertifikat
undefined
Water purification systems, Milli-Q® IQ 7003/05/10/15
Leverandør:  Merck
Spesifikasjoner
  • Catalog No:
  • 171-1379
  • 171-1378
  • 171-1380
  • 171-1381
  • Model:
  • Milli-Q® IQ 7005
  • Milli-Q® IQ 7003
  • Milli-Q® IQ 7010
  • Milli-Q® IQ 7015
  • Feed water:
  • Potable tap water (consult manual for requirements)
  • Potable tap water (consult manual for requirements)
  • Potable tap water (consult manual for requirements)
  • Potable tap water (consult manual for requirements)
  • Production flow rate:
  • 5 L/h
  • 3 L/h
  • 10 L/h
  • 15 L/h
  • TOC content:
  • ≤2 ppb (µg/L); typically ≤5 ppb (Type 1, from Q-POD® dispenser; in the appropriate operating conditions, otherwise typically ≤ 5 ppb.);
    ≤30 ppb (Type 2);
    (values are typical and may vary depending on the feed water)
  • ≤2 ppb (µg/L); typically ≤5 ppb (Type 1, from Q-POD® dispenser; in the appropriate operating conditions, otherwise typically ≤ 5 ppb.);
    ≤30 ppb (Type 2);
    (values are typical and may vary depending on the feed water)
  • ≤2 ppb (µg/L); typically ≤5 ppb (Type 1, from Q-POD® dispenser; in the appropriate operating conditions, otherwise typically ≤ 5 ppb.);
    ≤30 ppb (Type 2);
    (values are typical and may vary depending on the feed water)
  • ≤2 ppb (µg/L); typically ≤5 ppb (Type 1, from Q-POD® dispenser; in the appropriate operating conditions, otherwise typically ≤ 5 ppb.);
    ≤30 ppb (Type 2);
    (values are typical and may vary depending on the feed water)
  • Resistivity at +25 °C:
  • 18,2 MΩ·cm (Type 1);
    >5 MΩ·cm; typically 10 - 15 MΩ·cm (Type 2);
    (resistivity can also be displayed non-temperature-compensated as required by USP)
  • 18,2 MΩ·cm (Type 1);
    >5 MΩ·cm; typically 10 - 15 MΩ·cm (Type 2);
    (resistivity can also be displayed non-temperature-compensated as required by USP)
  • 18,2 MΩ·cm (Type 1);
    >5 MΩ·cm; typically 10 - 15 MΩ·cm (Type 2);
    (resistivity can also be displayed non-temperature-compensated as required by USP)
  • 18,2 MΩ·cm (Type 1);
    >5 MΩ·cm; typically 10 - 15 MΩ·cm (Type 2);
    (resistivity can also be displayed non-temperature-compensated as required by USP)
  • Bacteria:
  • <0,01 KBE/ml (Typ1 aus Q-POD®-Spender)
    <0,01 KBE/ml (Typ2 aus E-POD®-Spender+Endfilter)
    Mit Millipak®- oder Biopak®-Filter bei Installation + Verwendung in einer Laminar-Flow-Haube
    Werte sind typisch und können je nach Speisewasser variieren
  • <0,01 cfu/ml (Type 1, from Q-POD® dispenser);
    <0,01 cfu/ml (Type 2, from E-POD® dispenser with final filter);
    (With Millipak® or Biopak® filter when installed and used in a laminar flow hood; values are typical & may vary with the feed water)
  • <0,01 KBE/ml (Typ1 aus Q-POD®-Spender)
    <0,01 KBE/ml (Typ2 aus E-POD®-Spender+Endfilter)
    Mit Millipak®- oder Biopak®-Filter bei Installation + Verwendung in einer Laminar-Flow-Haube
    Werte sind typisch und können je nach Speisewasser variieren
  • <0,01 KBE/ml (Typ1 aus Q-POD®-Spender)
    <0,01 KBE/ml (Typ2 aus E-POD®-Spender+Endfilter)
    Mit Millipak®- oder Biopak®-Filter bei Installation + Verwendung in einer Laminar-Flow-Haube
    Werte sind typisch und können je nach Speisewasser variieren
  • Particles:
  • No particles with size >0,22 µm (Type 1, from Q-POD® dispenser);
    No particles with size >0.22 µm (Type 2, from E-POD® dispenser with final filter);
    (With Millipak® filter, with Biopak® polisher; values are typical and may vary depending on the nature and concentration of contaminants in the feed water)
  • No particles with size >0,22 µm (Type 1, from Q-POD® dispenser);
    No particles with size >0.22 µm (Type 2, from E-POD® dispenser with final filter);
    (With Millipak® filter, with Biopak® polisher; values are typical and may vary depending on the nature and concentration of contaminants in the feed water)
  • No particles with size >0,22 µm (Type 1, from Q-POD® dispenser);
    No particles with size >0.22 µm (Type 2, from E-POD® dispenser with final filter);
    (With Millipak® filter, with Biopak® polisher; values are typical and may vary depending on the nature and concentration of contaminants in the feed water)
  • No particles with size >0,22 µm (Type 1, from Q-POD® dispenser);
    No particles with size >0.22 µm (Type 2, from E-POD® dispenser with final filter);
    (With Millipak® filter, with Biopak® polisher; values are typical and may vary depending on the nature and concentration of contaminants in the feed water)
  • Endotoxin:
  • <0,001 EU/mL(Type 1, from Q-POD® dispenser);
    <0,001 EU/ml (Type 2, from an E-POD® dispenser with final filter);
    (With Biopak® polisher when installed and used in a laminar flow hood; values are typical and may vary depending on the nature and concentration of contaminants in the feed water)
  • <0,001 EU/mL(Type 1, from Q-POD® dispenser);
    <0,001 EU/ml (Type 2, from an E-POD® dispenser with final filter);
    (With Biopak® polisher when installed and used in a laminar flow hood; values are typical and may vary depending on the nature and concentration of contaminants in the feed water)
  • <0,001 EU/mL(Type 1, from Q-POD® dispenser);
    <0,001 EU/ml (Type 2, from an E-POD® dispenser with final filter);
    (With Biopak® polisher when installed and used in a laminar flow hood; values are typical and may vary depending on the nature and concentration of contaminants in the feed water)
  • <0,001 EU/mL(Type 1, from Q-POD® dispenser);
    <0,001 EU/ml (Type 2, from an E-POD® dispenser with final filter);
    (With Biopak® polisher when installed and used in a laminar flow hood; values are typical and may vary depending on the nature and concentration of contaminants in the feed water)
  • RNases:
  • <1 pg/ml (Type 1, from Q-POD® dispenser);
    <1 pg/ml (Type 2, from an E-POD® dispenser with final filter);
    (with Biopak® polisher; values are typical and may vary depending on the nature and concentration of contaminants in the feed water)
  • <1 pg/ml (Type 1, from Q-POD® dispenser);
    <1 pg/ml (Type 2, from an E-POD® dispenser with final filter);
    (with Biopak® polisher; values are typical and may vary depending on the nature and concentration of contaminants in the feed water)
  • <1 pg/ml (Type 1, from Q-POD® dispenser);
    <1 pg/ml (Type 2, from an E-POD® dispenser with final filter);
    (with Biopak® polisher; values are typical and may vary depending on the nature and concentration of contaminants in the feed water)
  • <1 pg/ml (Type 1, from Q-POD® dispenser);
    <1 pg/ml (Type 2, from an E-POD® dispenser with final filter);
    (with Biopak® polisher; values are typical and may vary depending on the nature and concentration of contaminants in the feed water)
  • DNases:
  • <5 pg/mL (Type 1, from Q-POD® dispenser)
    <5 pg/ml (Type 2, from an E-POD® dispenser with final filter)
    (With Biopak® polisher; values are typical & may vary with the feed water)
  • <5 pg/mL (Type 1, from Q-POD® dispenser)
    <5 pg/ml (Type 2, from an E-POD® dispenser with final filter)
    (With Biopak® polisher; values are typical & may vary with the feed water)
  • <5 pg/mL (Type 1, from Q-POD® dispenser)
    <5 pg/ml (Type 2, from an E-POD® dispenser with final filter)
    (With Biopak® polisher; values are typical & may vary with the feed water)
  • <5 pg/mL (Type 1, from Q-POD® dispenser)
    <5 pg/ml (Type 2, from an E-POD® dispenser with final filter)
    (With Biopak® polisher; values are typical & may vary with the feed water)
  • W×D×H:
  • 315×380×498 mm
  • 315×380×498 mm
  • 375×380×498 mm
  • 375×380×498 mm
  • Weight:
  • 26 kg
  • 26 kg
  • 29,7 kg
  • 30 kg
Spesifikasjoner
Consumables
Produktinformasjon