Order Entry
Norway
ContactUsLinkComponent
4-Aminophenol ≥98%, GPR RECTAPUR®
Health
Irritant
Environment
4-Aminophenol ≥98%, GPR RECTAPUR®
Catalog # 85913.230
Supplier:  VWR Chemicals
CAS Number:  
undefined
Health
Irritant
Environment
4-Aminophenol ≥98%, GPR RECTAPUR®
Catalog # 85913.230
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 g
  • Seq:
    3545

Specifications

Specification Test Results

  • Assay (GC) :
    ≥ 98.0 %
  • White to cream to beige to light brown:
    Passes test
  • Crystals or granules or powder:
    Passes test

Specification Test Results

About this item

 :
 :109.13 g/mol
Merck Index:13,00461
Boiling Pt:284 °C (1013 hPa)
Melting Pt:∼186 °C
Density:1.3 g/cm³ (20 °C)
Flash Pt:195 °C (closed cup)
EINECS:204-616-2
UN:2512
ADR:6.1,III
 :GHS08
 :GHS07
 :GHS09
Storage Temperature:Ambient
CAS Number:123-30-8
Formula:H₂NC₆H₄OH
MDL Number:MFCD00007869