Order Entry
Norway
ContactUsLinkComponent
Bromocresol green ≥95% (dye content), powder, AnalaR NORMAPUR® ACS pH-indicator
Bromocresol green ≥95% (dye content), powder, AnalaR NORMAPUR® ACS pH-indicator
Supplier:  VWR Chemicals
Certificates
undefined
Bromocresol green ≥95% (dye content), powder, AnalaR NORMAPUR® ACS pH-indicator
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Dye content (spectrophotometrically):
    ≥ 95 %
  • Appearance:
    Yellow to tan powder
  • Appearance of solution:
    Conforms
  • Identification (IR):
    Passes test
  • Sensitivity:
    Passes test
  • Transition range (according to ACS):
    Passes test
  • Visual transition interval:
    pH 3.8 - pH 5.4
  • Loss on drying (110°C):
    ≤ 3 %

Specification Test Results

Related Information