Order Entry
Norway
ContactUsLinkComponent
 
Travel trolley bag, B909
Catalog # PRTWB909BKR
Supplier:  Portwest
undefined
Travel trolley bag, B909
Catalog # PRTWB909BKR
Supplier:  Portwest

Specifications

  • Capacity:
    100 L
  • Colour:
    Black
  • Description:
    Travel trolley bag, holdalls
  • Pk:
    1

Specifications

About this item

Aesthetic styling and functional design make this travel trolley bag a must have item. Internally the bag is split into three sections with hook and loop strip dividers, making it ideal for separating your items.

  • Shell fabric: 100% polyester 600D fabric g
  • Contrast fabric: 210T PA g
  • ID card holder for ID card display
  • In-line wheel system
  • Telescopic handle
  • A tough and varied range of holdalls

A comprehensive collection of versatile luggage pieces, suitable for travelling or general everyday use. Robustly constructed with strong materials and tough components. Reliability and durability guarantee 100% customer satisfaction.