Analytical standard, Cyphenothrin, reference material, Fluka™
Catalog # HONCR1737-100MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mg
- Standards:Reference material
- CAS No.:39515-40-7
- Description:Cyphenothrin
- Description:Cyphenothrin
- Form:Neat