Analytical standard, Chlorthal-dimethyl, reference material, Fluka™
Catalog # HONCR1719-50MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:50 mg
- Standards:Reference material
- CAS No.:1861-32-1
- Description:Chlorthal-dimethyl
- Description:Chlorthal-dimethyl
- Form:Neat